Sodium hexadecyl sulfate ≥95.0% (by titrimetric analysis) (contains ca. 40% Sodium Stearyl Sulfate)

Supplier: TCI America

H0080-500G H0080-25G
TCH0080-500GEA 368.14 CAD
TCH0080-500G TCH0080-025G
Sodium hexadecyl sulfate ≥95.0% (by titrimetric analysis) (contains ca. 40% Sodium Stearyl Sulfate)
Sodium hexadecyl sulphate

CAS Number: 1120-01-0
MDL Number: MFCD00047766
Molecular Formula: C16H34O4S
Molecular Weight: 344.49
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: Cetylsulfuric Acid Sodium Salt, Sodium Cetyl Sulfate, Hexadecylsulfuric Acid Sodium Salt

Formula: C₁₆H₃₃NaO₄S
MDL Number: MFCD00047766
CAS Number: 1120-01-0
EINECS: 214-292-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9b06716867bc1e93f4a540c8caefecd7ccd6c44&vid=f2574532eb9555636ceff565859fa0784ca26003&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR