1-Hexadecanol ≥98.0%

Supplier: TCI America

H0071-500G H0071-25G
TCH0071-500GEA 102.56 CAD
TCH0071-500G TCH0071-025G
1-Hexadecanol ≥98.0%
1-Hexadecanol

CAS Number: 36653-82-4
MDL Number: MFCD00004760
Molecular Formula: C16H34O
Molecular Weight: 242.45
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 344
Melting point (°C): 50
Flash Point (°C): 175


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Cetanol, Cetyl Alcohol, Hexadecyl Alcohol

Formula: H₃C(CH₂)₁₄CH₂OH
Boiling Pt: 344 °C
Melting Pt: 50 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00004760
CAS Number: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2895c8007d4b4aed14c68b78929e771e2e962596&vid=0f94cb316dc8ab8ce2ccd3cc490368a201fd90e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR