1-Hexadecanethiol ≥97.0%

Supplier: TCI America

H0068-25ML H0068-500ML
TCH0068-25MLEA 58.46 CAD
TCH0068-25ML TCH0068-500ML
1-Hexadecanethiol ≥97.0%
1-Hexadecanethiol

CAS Number: 2917-26-2
MDL Number: MFCD00011677
Molecular Formula: C16H34S
Molecular Weight: 258.51
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 135
Freezing point (°C): 19
Specific Gravity (20/20): 0.85


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Cetanethiol, Cetyl Mercaptan, Hexadecyl Mercaptan

Formula: C₁₆H₃₄S
Boiling Pt: 189 °C
Melting Pt: 18…20 °C
Density: 0.85 (20 °C)
Flash Pt: 135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011677
CAS Number: 2917-26-2
EINECS: 220-846-6
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ca9c63c8de4f3b0f0d5eba82de60b8c6ba79f7b&vid=3c9dc6e9b3b84d2a27a6fd1146e4bf8cb7a94254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR