n-Hexadecane ≥98.0%

Supplier: TCI America
Danger

H0066-25ML H0066-500ML H0066-100ML
TCH0066-025MLEA 33.59 CAD
TCH0066-025ML TCH0066-500ML TCH0066-100ML
n-Hexadecane ≥98.0%
n-Hexadecane

CAS Number: 544-76-3
MDL Number: MFCD00008998
Molecular Formula: C16H34
Molecular Weight: 226.45
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 287
Flash Point (°C): 136
Freezing point (°C): 17
Specific Gravity (20/20): 0.77


Other Product Names / Synonyms: Cetane

Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C
Melting Pt: 18 °C
Density: 0.77 (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa2d185a1c6a13f018320f97f02b86d5ea0e1a73&vid=603479a5e5d4c0545025c9c12ded75dac6e42d7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR