(±)-2-Heptanol ≥98.0%

Supplier: TCI America

Synonyms: (±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol

H0034-25ML H0034-500ML H0034-100ML
TCH0034-025MLEA 36.34 CAD
TCH0034-025ML TCH0034-500ML TCH0034-100ML
(±)-2-Heptanol ≥98.0%
(±)-2-Heptanol

CAS Number: 543-49-7
MDL Number: MFCD00004587
Molecular Formula: C7H16O
Molecular Weight: 116.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 64
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Amylmethylcarbinol, 2-Heptyl Alcohol, Methylpentylcarbinol

Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 160
Density: 0.82 (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00004587
CAS Number: 543-49-7
EINECS: 208-844-3
UN: 1987
ADR: 3 III
Merck Index: 13,04682
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75de1740a4cbcae47b0a6b3c24ec691748f77592&vid=5ef2ad3205eb05dc648382669525ee49363ae0e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR