Ethyl enantate ≥98.0%

Supplier: TCI America

Synonyms: Heptylic acid ethyl ester, Enanthic acid ethyl ester, Heptanoic acid ethyl ester, Ethyl heptanoate

H0031-500ML H0031-25ML
TCH0031-500MLEA 148.52 CAD
TCH0031-500ML TCH0031-025ML
Ethyl enantate ≥98.0%
Ethyl enantate

CAS Number: 106-30-9
MDL Number: MFCD00009538
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Flash Point (°C): 71
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Enanthic Acid Ethyl Ester, Ethyl Enanthate, Heptanoic Acid Ethyl Ester

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 372
Melting Pt: -66
Density: 0.87 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00009538
CAS Number: 106-30-9
EINECS: 203-382-9
Merck Index: 13,03867
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02153c1499d70f35126367e28c25f2276557c8f3&vid=02153c1499d70f35126367e28c25f2276557c8f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR