n-Heptane ≥99.0%
Supplier: TCI America
Synonyms:
high purity solvents
H0027-100ML
TCH0027-100MLEA
77.45
CAD
TCH0027-100ML
n-Heptane ≥99.0%
n-Heptane
[for Spectrophotometry]
CAS Number: 142-82-5
MDL Number: MFCD00009544
Molecular Formula: C7H16
Molecular Weight: 100.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Melting point (°C): -91
Flash Point (°C): -2
Specific Gravity (20/20): 0.68
Formula:
H₃C(CH₂)₅CH₃ MW: 100.20 g/mol Boiling Pt: 98 °C Melting Pt: –91 °C Density: 0.68 (20 °C) Flash Pt: –2 °C |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 UN: 1206 ADR: 3,II Merck Index: 13,04679 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...