n-Heptadecane ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
N-heptadecane
H0023-25G
H0023-250G
TCH0023-025GEA
54.67
CAD
TCH0023-025G
TCH0023-250G
n-Heptadecane ≥99.0%
n-Heptadecane
CAS Number: 629-78-7
MDL Number: MFCD00009002
Molecular Formula: C17H36
Molecular Weight: 240.48
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 303
Flash Point (°C): 149
Freezing point (°C): 22
Formula:
H₃C(CH₂)₁₅CH₃ Boiling Pt: 303 °C Melting Pt: 21.2 °C Density: 0.7783 g/cm³ (20 °C) Flash Pt: 149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009002 CAS Number: 629-78-7 EINECS: 211-108-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...