1-Heptadecanol ≥97.0%
Supplier: TCI America
H0018-5G
H0018-25G
TCH0018-005GEA
129.56
CAD
TCH0018-005G
TCH0018-025G
1-Heptadecanol ≥97.0%
1-Heptadecanol
CAS Number: 1454-85-9
MDL Number: MFCD00002822
Molecular Formula: C17H36O
Molecular Weight: 256.47
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 309
Melting point (°C): 54
Flash Point (°C): 154
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Heptadecyl Alcohol
Formula:
H₃C(CH₂)₁₅CH₂OH Boiling Pt: 309 °C Melting Pt: 54 °C Density: 0.967 g/cm³ (20 °C) Flash Pt: 154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002822 CAS Number: 1454-85-9 EINECS: 215-932-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...