Hematoxylin hydrate ≥97.0% (by GC)

Supplier: TCI America

H0006-5G H0006-25G
TCH0006-5GEA 113.76 CAD
TCH0006-5G TCH0006-025G
Hematoxylin hydrate ≥97.0% (by GC)
Hematoxylin hydrate

CAS Number: 517-28-2
MDL Number: MFCD00078111
Molecular Formula: C16H14O6
Molecular Weight: 302.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal


Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: Natural Black 1

Formula: C₁₆H₁₄O₆·xH₂O
Melting Pt: 100…120 °C
MDL Number: MFCD00078111
CAS Number: 517-28-2
EINECS: 208-237-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a123e4739ba96a5ae0c41535943329e77cf2c705&vid=dbb37bf768fc06f4262b2372eade62039c8487a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR