L(-)-Galactose ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (2S,3R,4R,5S)-2,3,4,5,6-Pentahydroxyhexanal, (2S,3R,4R,5S)-Pentahydroxyhexanal, L-Galactose

G0267-500MG G0267-100MG
TCG0267-500MGEA 896.52 CAD
TCG0267-500MG TCG0267-100MG
L(-)-Galactose ≥98.0% (by HPLC)
L(-)-Galactose

CAS Number: 15572-79-9
MDL Number: MFCD00063833
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -79 deg (C=1, H2O)

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00063833
CAS Number: 15572-79-9
EINECS: 239-630-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6c87e8de740f38fe5842b13e4323760b414a7b6&vid=a6c87e8de740f38fe5842b13e4323760b414a7b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR