Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) 98,0
Supplier: TCI America
Synonyms:
H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl
G0254-25G
G0254-1G
G0254-5G
TCG0254-25GEA
117.32
CAD
TCG0254-25G
TCG0254-001G
TCG0254-005G
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) 98,0
Glycine tert-butyl ester hydrochloride (H-Gly-OtBu.HCl)
CAS Number: 27532-96-3
MDL Number: MFCD00058255
Molecular Formula: C6H13NO2
Molecular Weight: 167.63
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Gly-OtBu.HCl
Formula:
C₆H₁₄ClNO₂ MW: 167.64 g/mol |
MDL Number:
MFCD00058255 CAS Number: 27532-96-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...