Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) ≥97.0%

Supplier: TCI America

Synonyms: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl

G0246-25G G0246-500G
TCG0246-025GEA 38.9 CAD
TCG0246-025G TCG0246-500G
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) ≥97.0%
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)

CAS Number: 5680-79-5
MDL Number: MFCD00012870
Molecular Formula: C3H7NO2
Molecular Weight: 125.55
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 171


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Gly-OMe.HCl

Formula: C₃H₈ClNO₂
MW: 125.56 g/mol
Melting Pt: 171 °C
MDL Number: MFCD00012870
CAS Number: 5680-79-5
EINECS: 227-139-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3851f29652f700732f78f48520c63310c78aafa7&vid=3851f29652f700732f78f48520c63310c78aafa7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR