Guanidinium thiocyanate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Guanidinium sulfocyanide
, Guanidine Thiocyanate
, Guanidinium sulphocyanide
, GTC
, Guanidinium rhodanide
G0230-25G
G0230-500G
TCG0230-025GEA
58.95
CAD
TCG0230-025G
TCG0230-500G
Guanidinium thiocyanate ≥99.0% (by titrimetric analysis)
Guanidinium thiocyanate
CAS Number: 593-84-0
MDL Number: MFCD00013027
Molecular Formula: CH5N3
Molecular Weight: 118.16
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 120
Formula:
CH₅N₃·CHNS MW: 118.16 g/mol Melting Pt: 120 °C Density: 1.29 g/cm³ (20 °C) |
MDL Number:
MFCD00013027 CAS Number: 593-84-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...