L(-)-Glucose ≥98.0%

Supplier: TCI America

Synonyms: (2S,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal

G0226-5G G0226-1G
TCG0226-5GEA 606.72 CAD
TCG0226-5G TCG0226-001G
L(-)-Glucose ≥98.0%
L(-)-Glucose

CAS Number: 921-60-8
MDL Number: MFCD00148913
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Specific rotation [a]20/D: -52 deg (C=5, H2O)

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376
Melting Pt: 146…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00148913
CAS Number: 921-60-8
EINECS: 213-068-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d65657a21d35dfa849425e1d1c8b44688e6c28c3&vid=d65657a21d35dfa849425e1d1c8b44688e6c28c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR