(3-Glycidoxypropyl)trimethoxysilane ≥97.0%

Supplier: TCI America

G0210-500G G0210-25G G0210-100G
TCG0210-500GEA 195.65 CAD
TCG0210-500G TCG0210-025G TCG0210-100G
(3-Glycidoxypropyl)trimethoxysilane ≥97.0%
(3-Glycidoxypropyl)trimethoxysilane

CAS Number: 2530-83-8
MDL Number: MFCD00005144
Molecular Formula: C9H20O5Si
Molecular Weight: 236.34
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 137
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Silane Coupling Agents, Monomers
Other Product Names / Synonyms: Glycidyl 3-Trimethoxysilylpropyl Ether, GOPTS

Formula: C₉H₂₀O₅Si
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.07 (20 °C)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00005144
CAS Number: 2530-83-8
EINECS: 219-784-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5951c6723e40a733073c212a91745eca9bf548f8&vid=5951c6723e40a733073c212a91745eca9bf548f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR