Octyl-3,4,5-trihydroxybenzoate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Octyl 3,4,5-trihydroxybenzoate, Gallic acid octyl ester, Octyl gallate, n-Octyl gallate

G0206-250G G0206-25G
TCG0206-250GEA 372.88 CAD
TCG0206-250G TCG0206-025G
Octyl-3,4,5-trihydroxybenzoate ≥98.0% (by titrimetric analysis)
Octyl 3,4,5-trihydroxybenzoate

CAS Number: 1034-01-1
MDL Number: MFCD00002197
Molecular Formula: C15H22O5
Molecular Weight: 282.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: Gallic Acid n-Octyl Ester

Formula: C₁₅H₂₂O₅
MW: 282.34 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00002197
CAS Number: 1034-01-1
EINECS: 213-853-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b36c29337f140113485a6207c407478a7b63c1da&vid=b36c29337f140113485a6207c407478a7b63c1da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR