Methyl violet 2B

Supplier: TCI America
Danger

Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride

G0177-25G G0177-100G
TCG0177-025GEA 35.31 CAD
TCG0177-025G TCG0177-100G
Methyl violet 2B
Methyl Violet 2B

CAS Number: 8004-87-3
MDL Number: MFCD00012568
Form: Crystal
Color: Deep Yellow Green


Other Product Names / Synonyms: Aniline Violet, Dahlia Violet B, Methyl Violet

Formula: C₂₄H₂₈ClN₃
MW: 393.96 g/mol
Melting Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00012568
CAS Number: 8004-87-3
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR