Guanosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

G0171-5G G0171-25G
TCG0171-005GEA 33.18 CAD
TCG0171-005G TCG0171-025G
Guanosine ≥98.0% (by HPLC, titration analysis)
Guanosine

CAS Number: 118-00-3
MDL Number: MFCD00010182
Molecular Formula: C10H13N5O5
Molecular Weight: 283.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -76 deg (C=1, 1mol/L NaOH sol.)

Formula: C₁₀H₁₃N₅O₅
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS Number: 118-00-3
EINECS: 204-227-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4edb1fbea8e30e86e72cff16ab729385ce0d79f5&vid=faaf1c10e997b8023bf60922b414c967b74725cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR