Glycylglycylglycine (Gly-Gly-Gly)
Supplier: TCI America
G0126-5G
G0126-25G
TCG0126-005GEA
110.76
CAD
TCG0126-005G
TCG0126-025G
Glycylglycylglycine (Gly-Gly-Gly)
Proteins and Peptides
CAS Number: 556-33-2
MDL Number: MFCD00036223
Molecular Formula: C6H11N3O4
Molecular Weight: 189.17
Purity/Analysis Method:
- EINECS: 209-122-0
- Solubility in water: Almost transparency
- Product Categories: Bioscience, Oligopeptides
- Other Product Names/Synonyms: H-Gly-Gly-Gly-OH, Triglycine
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Gly-Gly-OH, Triglycine
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...