Glycylglycine (Gly-Gly)

Supplier: TCI America

G0124-25G G0124-500G G0124-100G
TCG0124-025GEA 80.06 CAD
TCG0124-025G TCG0124-500G TCG0124-100G
Glycylglycine (Gly-Gly)
Proteins and Peptides

Product Categories: Bioscience, Buffer-related Reagents, Oligopeptides
Other Product Names / Synonyms: H-Gly-Gly-OH, Diglycine


  • Melting point: 260 °C
  • GHS symbol and signal word: GHS07 and warning
  • EINECS: 209-127-8
  • Merck index: 14,04503
  • Solubility in water: Transparency
  • Drying loss: Max. 0.2%
  • Ignition residue (sulfate): Max. 0.05%


Product Categories: Bioscience, Buffer-related Reagents, Oligopeptides Other Product Names / Synonyms: H-Gly-Gly-OH, Diglycine

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05da448f499fa73f6edb1f95f5dbb94814a59af4&vid=098c51a29b12bfaf450c93b9a0a2ff1c044910a9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR