Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99.0% (by titrimetric analysis)
Supplier: TCI America
G0102-25G
G0102-500G
TCG0102-025GEA
28.95
CAD
TCG0102-025G
TCG0102-500G
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99.0% (by titrimetric analysis)
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)
CAS Number: 623-33-6
MDL Number: MFCD00012871
Molecular Formula: C4H9NO2
Molecular Weight: 139.58
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 146
Product Categories: Synthetic Organic Chemistry, Precursors for Preparation of Highly Reactive Reagents, Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Gly-OEt.HCl
Formula:
C₄H₁₀ClNO₂ Melting Pt: 146 °C |
MDL Number:
MFCD00012871 CAS Number: 623-33-6 EINECS: 210-787-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...