Glutaric anhydride ≥98.0%
Supplier: TCI America
Synonyms:
Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione
G0071-25G
G0071-500G
G0071-100G
TCG0071-025GEA
41.08
CAD
TCG0071-025G
TCG0071-500G
TCG0071-100G
Glutaric anhydride ≥98.0%
Glutaric anhydride
CAS Number: 108-55-4
MDL Number: MFCD00006679
Molecular Formula: C5H6O3
Molecular Weight: 114.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 146
Melting point (°C): 56
Flash Point (°C): 178
Formula:
C₅H₆O₃ MW: 114.1 g/mol Boiling Pt: 287 Melting Pt: 56 °C Density: 1.411 g/cm³ (20 °C) Flash Pt: 178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006679 CAS Number: 108-55-4 EINECS: 203-593-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...