Glutaric acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1,5-Pentanedioic acid

G0069-25G G0069-500G
TCG0069-025GEA 34.3 CAD
TCG0069-025G TCG0069-500G
Glutaric acid ≥99.0%
Glutaric acid

CAS Number: 110-94-1
MDL Number: MFCD00004410
Molecular Formula: C5H8O4
Molecular Weight: 132.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 98


Other Product Names / Synonyms: Pentanedioic Acid

Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 302
Melting Pt: 98
Density: 1.43 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004410
CAS Number: 110-94-1
EINECS: 203-817-2
Merck Index: 13,04486

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR