L(+)-Glutamine ≥99.0%

Supplier: TCI America

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

G0063-500G G0063-25G G0063-100G
TCG0063-500GEA 214.88 CAD
TCG0063-500G TCG0063-025G TCG0063-100G
L(+)-Glutamine ≥99.0%
L(+)-Glutamine

CAS Number: 56-85-9
MDL Number: MFCD00008044
Molecular Formula: C5H10N2O3
Molecular Weight: 146.15
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 186
Specific rotation [a]20/D: 6.8 deg (C=4, H2O)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Gln-OH

Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Boiling Pt: 355
Melting Pt: 186 °C
Density: 1.5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c15d9b50993943ae27d8bf552b85fda28dfecaa&vid=4c15d9b50993943ae27d8bf552b85fda28dfecaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR