(±)-2-Pyrrolidinone-5-carboxylic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

G0061-25G G0061-500G G0061-100G
TCG0061-025GEA 45.82 CAD
TCG0061-025G TCG0061-500G TCG0061-100G
(±)-2-Pyrrolidinone-5-carboxylic acid ≥99.0% (by titrimetric analysis)
(±)-2-Pyrrolidinone-5-carboxylic acid

CAS Number: 149-87-1
MDL Number: MFCD00064322
Molecular Formula: C5H7NO3
Molecular Weight: 129.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 184


Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: DL-Glutamic Acid Lactam, 2-Pyrrolidone-5-carboxylic Acid, 5-Oxopyrrolidine-2-carboxylic Acid, H-DL-Pyr-OH

Formula: C₅H₇NO₃
Melting Pt: 184 °C
MDL Number: MFCD00064322
CAS Number: 149-87-1
EINECS: 205-748-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f46c78182a093a46c28ad6a3acfe90067f0c37c&vid=1f46c78182a093a46c28ad6a3acfe90067f0c37c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR