D-(+)-Glucose ≥98.0%

Supplier: TCI America

Synonyms: Corn sugarD-GlucoseD-(+)-DextroseGlucolinᴅ-(+)-Glucose(2R,3S,4R,5R)-2,3,4,5,6-PentahydroxyhexanalGlucoseDextrose

G0048-500G G0048-25G
TCG0048-500GEA 42.52 CAD
TCG0048-500G TCG0048-25G
D-(+)-Glucose ≥98.0%
D(+)-Glucose

CAS Number: 50-99-7
MDL Number: MFCD00148912
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: 53 deg (C=10, H2O)


Product Categories: Bioscience, Basic Sugars (Mono & Oligosaccharides), Glucose
Other Product Names / Synonyms: D-(+)-Dextrose

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146 °C
Density: 1.54 g/cm³ (20 °C)
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459
https://ca.vwr.com/stibo/web/std.lang.all/97/43/28969743.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR