Sodium gluconate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

G0041-500G G0041-25G
TCG0041-500GEA 66.06 CAD
TCG0041-500G TCG0041-025G
Sodium gluconate ≥99.0% (by titrimetric analysis)
Sodium gluconate

CAS Number: 527-07-1
MDL Number: MFCD00064210
Molecular Formula: C6H12O7
Molecular Weight: 218.14
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 206
Specific rotation [a]20/D: 13 deg (C=2, H2O)


Product Categories: Bioscience, Sugar Acids, Glucose
Other Product Names / Synonyms: Gluconic Acid Sodium Salt

Formula: HOH₂C(CH(OH))₄COONa
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e896b77f0f7bb9db09339d111b36e5f7d11a34e2&vid=e896b77f0f7bb9db09339d111b36e5f7d11a34e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR