D-Glucono-1,5-lactone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone

G0039-25G G0039-500G
TCG0039-025GEA 34.24 CAD
TCG0039-025G TCG0039-500G
D-Glucono-1,5-lactone ≥98.0% (by titrimetric analysis)
D-Glucono-1,5-lactone

CAS Number: 90-80-2
MDL Number: MFCD00006647
Molecular Formula: C6H10O6
Molecular Weight: 178.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 153
Specific rotation [a]20/D: 63.5 deg (C=10, H2O)


Product Categories: Bioscience, Sugar Acids, Glucose
Other Product Names / Synonyms: Gluconic Acid Anhydride, Gluconic Acid delta-Lactone

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 385 °C (1013 hPa)
Melting Pt: 153 °C
Density: 1.61 g/cm³ (–5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006647
CAS Number: 90-80-2
EINECS: 202-016-5
Merck Index: 13,04470
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c71f1736ada2f0dbb12feee0d74a4d066142a0b6&vid=c71f1736ada2f0dbb12feee0d74a4d066142a0b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR