Calcium-D-gluconate monohydrate ≥98.0%

Supplier: TCI America

Synonyms: D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate

G0037-25G G0037-500G
TCG0037-025GEA 41.81 CAD
TCG0037-025G TCG0037-500G
Calcium-D-gluconate monohydrate ≥98.0%
Calcium D-gluconate monohydrate

CAS Number: 299-28-5
MDL Number: MFCD00064209
Molecular Formula: C6H12O7
Molecular Weight: 430.37
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, Sugar Acids, Glucose
Other Product Names / Synonyms: Gluconic Acid Calcium Salt

Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Melting Pt: 178
Storage Temperature: Ambient
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df3245f5acabaa949b5e458cda401178d1efac7a&vid=df3245f5acabaa949b5e458cda401178d1efac7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR