Geraniol ≥96.0%

Supplier: TCI America
Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

G0027-25ML G0027-500ML G0027-100ML
TCG0027-025MLEA 45.91 CAD
TCG0027-025ML TCG0027-500ML TCG0027-100ML
Geraniol ≥96.0%
Geraniol

CAS Number: 106-24-1
MDL Number: MFCD00002917
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Melting point (°C): -15
Flash Point (°C): 112
Specific Gravity (20/20): 0.88


Product Categories: Bioscience, Acyclic Monoterpenes
Other Product Names / Synonyms: 3,7-Dimethyl-2,6-octadien-1-ol

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 230 °C
Melting Pt: –15 °C
Density: 0.88 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb7e8a01e1abdd5ba4d441c44c806a4bc8d9a898&vid=bb7e8a01e1abdd5ba4d441c44c806a4bc8d9a898&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR