Methyl-3,4,5-trihydroxybenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

G0017-500G G0017-25G G0017-100G
TCG0017-500GEA 350.76 CAD
TCG0017-500G TCG0017-025G TCG0017-100G
Methyl-3,4,5-trihydroxybenzoate ≥98.0% (by GC, titration analysis)
Methyl 3,4,5-trihydroxybenzoate

CAS Number: 99-24-1
MDL Number: MFCD00002194
Molecular Formula: C8H8O5
Molecular Weight: 184.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 200


Other Product Names / Synonyms: Gallic Acid Methyl Ester

Formula: C₈H₈O₅
Boiling Pt: 450 °C (1013 hPa)
Melting Pt: 200 °C
MDL Number: MFCD00002194
CAS Number: 99-24-1
EINECS: 202-741-7
Merck Index: 13,04366
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dcd980fd897e3c6309d5917ba0c366ac74b0f2a&vid=5dcd980fd897e3c6309d5917ba0c366ac74b0f2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR