Ethyl-3,4,5-trihydroxybenzoate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0016-25G G0016-500G G0016-100G
TCG0016-025GEA 45.82 CAD
TCG0016-025G TCG0016-500G TCG0016-100G
Ethyl-3,4,5-trihydroxybenzoate ≥98.0% (by titrimetric analysis)
Ethyl 3,4,5-trihydroxybenzoate

CAS Number: 831-61-8
MDL Number: MFCD00016430
Molecular Formula: C9H10O5
Molecular Weight: 198.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 153


Other Product Names / Synonyms: Gallic Acid Ethyl Ester

Formula: C₉H₁₀O₅
Melting Pt: 153 °C
MDL Number: MFCD00016430
CAS Number: 831-61-8
EINECS: 212-608-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5624b856f6aa0dbc5ac9adb4abfd863b42924fd9&vid=5624b856f6aa0dbc5ac9adb4abfd863b42924fd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR