Dodecyl-3,4,5-trihydroxybenzoate

Supplier: TCI America
Warning

Synonyms: Gallic acid lauryl ester, n-Dodecyl gallate, Dodecyl 3,4,5-trihydroxybenzoate, Gallic acid dodecyl ester, Dodecyl gallate, Lauryl 3,4,5-trihydroxybenzoate, Lauryl gallate, Progalline LA

G0015-25G G0015-500G
TCG0015-025GEA 49.11 CAD
TCG0015-025G TCG0015-500G
Dodecyl-3,4,5-trihydroxybenzoate
Dodecyl 3,4,5-trihydroxybenzoate

CAS Number: 1166-52-5
MDL Number: MFCD00002195
Molecular Formula: C19H30O5
Molecular Weight: 338.44
Form: Crystal
Color: White
Melting point (°C): 96


Other Product Names / Synonyms: Gallic Acid Dodecyl Ester, Gallic Acid Lauryl Ester, Lauryl Gallate

Formula: C₁₉H₃₀O₅
Melting Pt: 96 °C
MDL Number: MFCD00002195
CAS Number: 1166-52-5
EINECS: 214-620-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1c5306be1f6342284370b297a279647374de7da&vid=c1c5306be1f6342284370b297a279647374de7da&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR