Gallic acid monohydrate ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate

G0011-500G G0011-25G G0011-100G
TCG0011-500GEA 189.6 CAD
TCG0011-500G TCG0011-025G TCG0011-100G
Gallic acid monohydrate ≥98.0%
Gallic acid monohydrate

CAS Number: 5995-86-8
MDL Number: MFCD00149098
Molecular Formula: C7H6O5
Molecular Weight: 170.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 253
Flash Point (°C): 250


Product Categories: Bioscience, Antioxidants, Others (Phytochemicals)
Other Product Names / Synonyms: 3,4,5-Trihydroxybenzoic Acid

Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: 250
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8
EINECS: 205-749-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a67d26092bafc1ee8ddb646cc6afe08c5b5235d&vid=1a67d26092bafc1ee8ddb646cc6afe08c5b5235d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR