Galactitol ≥98.0%
Supplier: TCI America
Synonyms:
Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
G0005-25G
G0005-250G
TCG0005-025GEA
113.93
CAD
TCG0005-025G
TCG0005-250G
Galactitol ≥98.0%
Galactitol
CAS Number: 608-66-2
MDL Number: MFCD00064288
Molecular Formula: C6H14O6
Molecular Weight: 182.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 280
Melting point (°C): 189
Product Categories: Bioscience, Sugar Alcohols, Galactose
Other Product Names / Synonyms: Dulcitol
Formula:
HOH₂C(CH(OH))₄CH₂OH Boiling Pt: 280 °C Melting Pt: 189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064288 CAS Number: 608-66-2 EINECS: 210-165-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...