2'-Fluoro-4'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
F0418-1G
F0418-5G
TCF0418-001GEA
48.98
CAD
TCF0418-001G
TCF0418-005G
2'-Fluoro-4'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Fluoro-4'-hydroxyacetophenone
CAS Number: 98619-07-9
MDL Number: MFCD00142713
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125
Flash Point (°C): 98
Formula:
C₈H₇FO₂ Melting Pt: 125 °C Flash Pt: 98 °C |
MDL Number:
MFCD00142713 CAS Number: 98619-07-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...