3-Fluoro-4-hydroxybenzoic acid, hydrate ≥98.0%
Supplier: TCI America
F0413-1G
F0413-5G
TCF0413-1GEA
66.36
CAD
TCF0413-1G
TCF0413-005G
3-Fluoro-4-hydroxybenzoic acid, hydrate ≥98.0%
3-Fluoro-4-hydroxybenzoic acid
CAS Number: 350-29-8
MDL Number: MFCD02094741
Molecular Formula: C7H5FO3
Molecular Weight: 156.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 163
Formula:
C₇H₅FO₃ MW: 156.11 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00143161 CAS Number: 350-29-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...