4-Fluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

F0405-25G F0405-5G F0405-1G
TCF0405-25GEA 282.82 CAD
TCF0405-25G TCF0405-5G TCF0405-001G
4-Fluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)
4-Fluoroanthranilic acid

CAS Number: 446-32-2
MDL Number: MFCD00075553
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-4-fluorobenzoic Acid

Formula: C₇H₆FNO₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 198…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00075553
CAS Number: 446-32-2
EINECS: 207-163-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20716acc8f8b346ab34bdabff9c0a09fbbc97434&vid=dcad2c45d1ed4a3803a39d98183d6a5e844f41cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR