Methyl-4-fluorobenzoate ≥97.0%

Supplier: TCI America

F0403-25G F0403-500G
TCF0403-25GEA 53.72 CAD
TCF0403-25G TCF0403-500G
Methyl-4-fluorobenzoate ≥97.0%
Methyl 4-fluorobenzoate

CAS Number: 403-33-8
MDL Number: MFCD00017959
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Flash Point (°C): 77
Specific Gravity (20/20): 1.20


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluorobenzoic Acid Methyl Ester

Formula: C₈H₇FO₂
Boiling Pt: 197 °C
Density: 1.20 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00017959
CAS Number: 403-33-8
EINECS: 206-956-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3c7222b51a5d74565be0d1e2ac1b883c6f9126e&vid=f3c7222b51a5d74565be0d1e2ac1b883c6f9126e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR