4-Fluoro-m-anisaldehyde ≥96.0%

Supplier: TCI America

F0401-5G F0401-25G F0401-1G
TCF0401-005GEA 131.14 CAD
TCF0401-005G TCF0401-25G TCF0401-1G
4-Fluoro-m-anisaldehyde ≥96.0%
4-Fluoro-m-anisaldehyde

CAS Number: 128495-46-5
MDL Number: MFCD00143320
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 60


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-3-methoxybenzaldehyde

Formula: C₈H₇FO₂
Melting Pt: 60 °C
MDL Number: MFCD00143320
CAS Number: 128495-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06273e0bef5cb3f3c31eb6c174419094b615210e&vid=06273e0bef5cb3f3c31eb6c174419094b615210e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR