4-Fluoro-3-nitrobenzoic acid ≥98.0%

Supplier: TCI America
Warning

F0399-25G F0399-5G
TCF0399-25GEA 197.5 CAD
TCF0399-25G TCF0399-005G
4-Fluoro-3-nitrobenzoic acid ≥98.0%
4-Fluoro-3-nitrobenzoic acid

CAS Number: 453-71-4
MDL Number: MFCD00007058
Molecular Formula: C7H4FNO4
Molecular Weight: 185.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 121

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Boiling Pt: 92
Melting Pt: 121 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=570cca0830bf66b0e1681a6c64b6a17dd7c04b30&vid=633cf7758b001ab9a0cb5842b0b14a6b9dc3fdd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR