5-Fluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
F0396-1G
F0396-5G
TCF0396-001GEA
71.1
CAD
TCF0396-001G
TCF0396-5G
5-Fluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)
5-Fluoroanthranilic acid
CAS Number: 446-08-2
MDL Number: MFCD00055566
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-5-fluorobenzoic Acid
Formula:
C₇H₆FNO₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055566 CAS Number: 446-08-2 EINECS: 207-159-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...