Ethyl-2-fluorobenzoate ≥98.0%

Supplier: TCI America

F0395-5G F0395-25G
TCF0395-005GEA 109.02 CAD
TCF0395-005G TCF0395-025G
Ethyl-2-fluorobenzoate ≥98.0%
Ethyl 2-fluorobenzoate

CAS Number: 443-26-5
MDL Number: MFCD00039214
Molecular Formula: C9H9FO2
Molecular Weight: 168.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Specific Gravity (20/20): 1.15


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluorobenzoic Acid Ethyl Ester

Formula: C₉H₉FO₂
Boiling Pt: 221 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00039214
CAS Number: 443-26-5
EINECS: 207-134-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=027f3e8b599547b2af8d4be43f3538b476386efc&vid=027f3e8b599547b2af8d4be43f3538b476386efc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR