Furan-2-boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
F0394-5G
F0394-1G
TCF0394-005GEA
132.72
CAD
TCF0394-005G
TCF0394-001G
Furan-2-boronic acid (contains varying amounts of Anhydride)
Furan-2-boronic acid
CAS Number: 13331-23-2
MDL Number: MFCD00799544
Molecular Formula: C4H5BO3
Molecular Weight: 111.89
Form: Crystal
Color: White
Storage Temperature: <0°C
Other Product Names / Synonyms: Furan-2-boronic Acid, 2-Furanylboronic Acid
Formula:
C₄H₅BO₃ |
MDL Number:
MFCD00799544 CAS Number: 13331-23-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...