2-Fluoro-5-nitrobenzoic acid ≥98.0%

Supplier: TCI America
Warning

F0391-25G F0391-5G
TCF0391-25GEA 319.16 CAD
TCF0391-25G TCF0391-005G
2-Fluoro-5-nitrobenzoic acid ≥98.0%
2-Fluoro-5-nitrobenzoic acid

CAS Number: 7304-32-7
MDL Number: MFCD00134238
Molecular Formula: C7H4FNO4
Molecular Weight: 185.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 139

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 139 °C
MDL Number: MFCD00134238
CAS Number: 7304-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2db40f626c46ecb10b3ce54347d0f04275d1bbe3&vid=2db40f626c46ecb10b3ce54347d0f04275d1bbe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR