2-Fluoro-p-anisaldehyde ≥96.0%

Supplier: TCI America

Synonyms: 2-Fluoro-4-methoxybenzaldehyde, 2-Fluoro-4-anisaldehyde

F0386-25G F0386-5G
TCF0386-25GEA 285.98 CAD
TCF0386-25G TCF0386-005G
2-Fluoro-p-anisaldehyde ≥96.0%
2-Fluoro-p-anisaldehyde

CAS Number: 331-64-6
MDL Number: MFCD00236679
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-4-methoxybenzaldehyde

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00236679
CAS Number: 331-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80d65b05934743c44a51e1cc8eae6fda6e02c417&vid=80d65b05934743c44a51e1cc8eae6fda6e02c417&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR