6-Fluoro-m-anisaldehyde ≥95.0%

Supplier: TCI America

Synonyms: 2-Fluoro-5-methoxybenzaldehyde, 6-Fluoro-3-anisaldehyde

F0383-5G F0383-25G
TCF0383-005GEA 263.86 CAD
TCF0383-005G TCF0383-25G
6-Fluoro-m-anisaldehyde ≥95.0%
6-Fluoro-m-anisaldehyde

CAS Number: 105728-90-3
MDL Number: MFCD00070795
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 230
Freezing point (°C): 23


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-5-methoxybenzaldehyde

Formula: C₈H₇FO₂
Boiling Pt: 230 °C
MDL Number: MFCD00070795
CAS Number: 105728-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2949d240a452af5a40cd2d468fdd32bc10808382&vid=2949d240a452af5a40cd2d468fdd32bc10808382&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR