4-Fluoro-2-nitroaniline ≥98.0%

Supplier: TCI America

F0380-25G F0380-5G
TCF0380-025GEA 284.24 CAD
TCF0380-025G TCF0380-005G
4-Fluoro-2-nitroaniline ≥98.0%
4-Fluoro-2-nitroaniline

CAS Number: 364-78-3
MDL Number: MFCD00007830
Molecular Formula: C6H5FN2O2
Molecular Weight: 156.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 94
Flash Point (°C): 89

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 95
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00007830
CAS Number: 364-78-3
EINECS: 206-666-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e427ae27e6b33d5c9ad7efecf5ec18b683799c6&vid=cd96268901b3894395ea5b10fe4fb85b516c9675&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR