5-Fluoro-1-indanone ≥96.0%

Supplier: TCI America

F0376-5G F0376-1G
TCF0376-5GEA 201.61 CAD
TCF0376-5G TCF0376-001G
5-Fluoro-1-indanone ≥96.0%
5-Fluoro-1-indanone

CAS Number: 700-84-5
MDL Number: MFCD00041031
Molecular Formula: C9H7FO
Molecular Weight: 150.15
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 37
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₉H₇FO
Boiling Pt: 114 °C
Melting Pt: 37 °C
Density: 1.35 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041031
CAS Number: 700-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ff810d3d51ceb8d33e5b691efbd85fa9ad30ab7&vid=4ff810d3d51ceb8d33e5b691efbd85fa9ad30ab7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR